missing translation for 'onlineSavingsMsg'
Learn More

Heptafluoro-2-iodopropane, 97%, stab. with copper powder

Product Code. 11399215
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399215 100 g 100g
11379215 5 g 5g
11389215 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11399215 Supplier Thermo Scientific Alfa Aesar Supplier No. L16523.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 677-69-0 | C3F7I | 295.926 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 677-69-0
Molecular Formula C3F7I
Molecular Weight (g/mol) 295.926
MDL Number MFCD00042089
InChI Key BBZVTTKMXRPMHZ-UHFFFAOYSA-N
Synonym heptafluoroisopropyl iodide, perfluoroisopropyl iodide, heptafluoro-2-iodopropane, 2-iodoheptafluoropropane, propane, 1,1,1,2,3,3,3-heptafluoro-2-iodo, 2-iodoperfluoropropane, propane, heptafluoro-2-iodo, perfluoro-2-iodopropane, heptafluoroisopropyliodide, 2-heptafluoroisopropyliodide
PubChem CID 33978
IUPAC Name 1,1,1,2,3,3,3-heptafluoro-2-iodopropane
SMILES C(C(F)(F)F)(C(F)(F)F)(F)I

Specifications

Melting Point -56°C
Density 2.1
Boiling Point 38°C
Linear Formula (CF3)2CFI
Refractive Index 1.33
Quantity 100 g
Beilstein 1841228
Sensitivity Light sensitive
Formula Weight 295.93
Percent Purity 97%
Chemical Name or Material Heptafluoro-2-iodopropane, stabilized with copper powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.