accessibility menu, dialog, popup

UserName

NHNOHO

CAS RN 677306-38-6

IUPAC Name: 1H-indazole-4-carboxylic acid
Synonyms: 1H-indazole-4-carboxylic acid
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: KGKZHHIUOZGUNP-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=NNC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results