missing translation for 'onlineSavingsMsg'
Learn More

1H-Indazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10749068 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10749068 1 g 1g
10562898 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10749068 Supplier Thermo Scientific Acros Supplier No. 430090010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 677306-38-6
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD03840644
InChI Key KGKZHHIUOZGUNP-UHFFFAOYSA-N
Synonym indazole-4-carboxylic acid, 1h-indazole-4-carboxylicacid, 4-indazolecarboxylic acid, 4-carboxy-1h-indazole, 2h-indazole-4-carboxylic acid, pubchem19651, acmc-1b3os, ksc352q1d, 1h-indazole-4-carboxylic acid, 4-1h indazole carboxylic acid
PubChem CID 21982323
IUPAC Name 1H-indazole-4-carboxylic acid
SMILES C1=CC(=C2C=NNC2=C1)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 209.5°C to 211.0°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 162.15
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1H-Indazole-4-carboxylic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1H-Indazole-4-carboxylic acid, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.