accessibility menu, dialog, popup

UserName

OOO

CAS RN 67869-90-3

IUPAC Name: 3,4-dihydro-2H-1,5-benzodioxepine-7-carbaldehyde
Synonyms: 3,4-dihydro-2H-1,5-benzodioxepine-7-carbaldehyde
Molecular Weight (g/mol): 178.19
Molecular Formula: C10H10O3
InChi Key: LCSVYSVGXQQHSI-UHFFFAOYSA-N
SMILES: O=CC1=CC2=C(OCCCO2)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)