missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepine-7-carbaldehyde, 95+%, Thermo Scientific™

Product Code. 10652635
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10652635 5 g Amber Glass Bottle 5g
10384562 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10652635 Supplier Thermo Scientific Maybridge Supplier No. CC13204DE
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 67869-90-3
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
InChI Key LCSVYSVGXQQHSI-UHFFFAOYSA-N
PubChem CID 2776388
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepine-7-carbaldehyde
SMILES C1COC2=C(C=C(C=C2)C=O)OC1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 2h-1,5-benzodioxepin-7-carboxaldehyde, 3,4-dihydro, 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carbaldehyde, 3,4-dihydro-2h-1,5-benzodioxepin-7-carboxaldehyde, 2h-1,5-benzodioxepin-7-carboxaldehyde,3,4-dihydro, 2h,3h,4h-benzo b 1,4-dioxepane-7-carbaldehyde, 3,4-dihydro-2h-1,5-benzodioxepine-7-carboxaldehyde, 3,4-dihydro-2h-benzo b 1,4 dioxepine-7-carbaldehyd
Formula Weight 178.19
Percent Purity 95%
Chemical Name or Material 3, 4-Dihydro-2H-1, 5-benzodioxepine-7-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.