accessibility menu, dialog, popup

UserName

BiH3CCH3OOH3CCH3OOH3CCH3OO

CAS RN 67874-71-9

IUPAC Name: bismuth(3+) tris(2-ethylhexanoate)
Synonyms: bismuth(3+) tris(2-ethylhexanoate)
Molecular Weight (g/mol): 638.6
Molecular Formula: C24H45BiO6
InChi Key: NUMHJBONQMZPBW-UHFFFAOYNA-K
SMILES: [Bi+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
Physical Form 
  • (6)
  • (3)
Quantity 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results