missing translation for 'onlineSavingsMsg'
Learn More

Bismuth 2-ethylhexanoate acid

Product Code. 15496965
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496965 5 g 5g
15406975 25 g 25g
15416975 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15496965 Supplier Thermo Scientific Alfa Aesar Supplier No. 045500.06
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 67874-71-9 | C24H48BiO6 | 641.622 g/mol

Bismuth 2-ethylhexanoate is used as an industrial lubricant, dessicant, and polyurethane catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67874-71-9
Molecular Formula C24H48BiO6
Molecular Weight (g/mol) 641.622
MDL Number MFCD00043803
InChI Key MWXWNYHYPMRCHN-UHFFFAOYSA-N
Synonym 2-Ethylhexanoic acid bismuth salt
PubChem CID 131857451
IUPAC Name bismuth;2-ethylhexanoic acid
SMILES CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.CCCCC(CC)C(=O)O.[Bi]

Specifications

Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 638.61
Percent Purity 92% (typically)
Physical Form Liquid
Chemical Name or Material Bismuth 2-ethylhexanoate, In 2-ethylhexanoic acid
compliance-icons
Product Identifier
  • Bismuth 2-ethylhexanoate, typically 92%, in 2-ethylhexanoic acid
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.