accessibility menu, dialog, popup

UserName

CAS RN 67899-41-6

FFFFFFFFFF

CAS RN 67899-41-6

IUPAC Name: 1,2,3,4,5-pentafluoro-6-(1,1,2,3,3-pentafluoroprop-2-en-1-yl)benzene
Synonyms: 1,2,3,4,5-pentafluoro-6-(1,1,2,3,3-pentafluoroprop-2-en-1-yl)benzene
Molecular Weight (g/mol): 298.08
Molecular Formula: C9F10
InChi Key: WRHBYJDZKRNITP-UHFFFAOYSA-N
SMILES: FC(F)=C(F)C(F)(F)C1=C(F)C(F)=C(F)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results