accessibility menu, dialog, popup

UserName

CH3OOHONH2

CAS RN 67973-80-2

IUPAC Name: methyl 3-amino-5-hydroxybenzoate
Synonyms: methyl 3-amino-5-hydroxybenzoate
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: DMNGQQIFOZYIRA-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(N)=CC(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results