accessibility menu, dialog, popup

UserName

CAS RN 681812-07-7

BFFFFFFHOOH

CAS RN 681812-07-7

IUPAC Name: [2,6-bis(trifluoromethyl)phenyl]boronic acid
Synonyms: 2,6-bis(trifluoromethyl)phenylboronic acid
Molecular Weight (g/mol): 257.93
Molecular Formula: C8H5BF6O2
InChi Key: WAMPGNNEOZGBHR-UHFFFAOYSA-N
SMILES: OB(O)C1=C(C=CC=C1C(F)(F)F)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results