accessibility menu, dialog, popup

UserName

CAS RN 6833-15-4

IUPAC Name: 4-chloro-N-phenylbenzamide
Synonyms: 4-chloro-N-phenylbenzamide
Molecular Weight (g/mol): 231.68
Molecular Formula: C13H10ClNO
InChi Key: SFHDVPIEJXCMBP-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results