accessibility menu, dialog, popup

UserName

CAS RN 6833-18-7

CH3ONH

CAS RN 6833-18-7

IUPAC Name: 4-methyl-N-phenylbenzamide
Synonyms: 4-methyl-N-phenylbenzamide
Molecular Weight (g/mol): 211.26
Molecular Formula: C14H13NO
InChi Key: PQEOPHYIUYAVDQ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results