accessibility menu, dialog, popup

UserName

OOOCl

CAS RN 6845-81-4

IUPAC Name: 2-(2H-1,3-benzodioxol-5-yl)acetyl chloride
Synonyms: 2-(2H-1,3-benzodioxol-5-yl)acetyl chloride
Molecular Weight (g/mol): 198.6
Molecular Formula: C9H7ClO3
InChi Key: HGSZGCXMAJSUSC-UHFFFAOYSA-N
SMILES: ClC(=O)CC1=CC2=C(OCO2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results