missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxol-5-ylacetyl chloride, ≥97%, Thermo Scientific™

Product Code. 10422844
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10422844 250 mg 250mg
10245624 1 g 1g
10255624 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10422844 Supplier Thermo Scientific Maybridge Supplier No. CC50802CB
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 6845-81-4
Molecular Formula C9H7ClO3
Molecular Weight (g/mol) 198.60
MDL Number MFCD03424722
InChI Key HGSZGCXMAJSUSC-UHFFFAOYSA-N
Synonym benzo 1,3 dioxol-5-yl-acetyl chloride, 1,3-benzodioxol-5-ylacetyl chloride, 2-benzo d 1,3 dioxol-5-yl acetyl chloride, benzo 1,3 dioxol-5-ylacetyl chloride, 2-2h-1,3-benzodioxol-5-yl acetyl chloride, 2-2h-benzo 3,4-d 1,3-dioxolen-5-yl acetyl chloride, 1,3-benzodioxole-5-acetylchloride, 3,4-methylenedioxyphenylacetylchloride, 3,4-methylenedioxyphenylacetyl chloride
PubChem CID 2756584
SMILES ClC(=O)CC1=CC2=C(OCO2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
IUPAC Name 2-(1,3-benzodioxol-5-yl)acetyl chloride
Formula Weight 198.6
Percent Purity 97%
Chemical Name or Material 1, 3-Benzodioxol-5-ylacetyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.