accessibility menu, dialog, popup

UserName

NH2NSSS

CAS RN 6846-35-1

IUPAC Name: 5-amino-3H-1,2,4-dithiazole-3-thione
Synonyms: 5-amino-1,2,4-dithiazole-3-thione
Molecular Weight (g/mol): 150.23
Molecular Formula: C2H2N2S3
InChi Key: YWZHEXZIISFIDA-UHFFFAOYSA-N
SMILES: NC1=NC(=S)SS1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results