missing translation for 'onlineSavingsMsg'
Learn More

Xanthane hydride, Thermo Scientific Chemicals

Product Code. 17251196 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17251196 25 g 25g
17241196 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17251196 Supplier Thermo Scientific Acros Supplier No. 467340250
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 6846-35-1 | C2H2N2S3 | 150.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 6846-35-1
Molecular Formula C2H2N2S3
Molecular Weight (g/mol) 150.23
InChI Key YWZHEXZIISFIDA-UHFFFAOYSA-N
IUPAC Name 5-amino-3H-1,2,4-dithiazole-3-thione
SMILES NC1=NC(=S)SS1

Spezifikation

Melting Point 145.0°C to 150.0°C
Quantity 25 g
Formula Weight 150.22
Chemical Name or Material Xanthane hydride
compliance-icons
Produktbeteckning
  • Xanthane hydride
Signalord
  • Warning
Riskkategorier
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faroangivelser
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Kompletterande information
  • MIXTURE LIST-Contain: 5-Amino-3H-1,2,4-dithiazole-3-thione

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.