accessibility menu, dialog, popup

UserName

A(a)

CAS RN 68478-92-2

IUPAC Name: ethenyl[(ethenyldimethylsilyl)oxy]dimethylsilane platinum
Synonyms: ethenyl[(ethenyldimethylsilyl)oxy]dimethylsilane platinum
Molecular Weight (g/mol): 381.49
Molecular Formula: C8H18OPtSi2
InChi Key: RCNRJBWHLARWRP-UHFFFAOYSA-N
SMILES: [Pt].C[Si](C)(O[Si](C)(C)C=C)C=C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)