missing translation for 'onlineSavingsMsg'
Learn More

Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, soln. in vinyl terminated polydimethylsiloxane

Product Code. 11324258
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324258 5 g 5g
11314258 1 g 1g
11304258 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324258 Supplier Thermo Scientific Alfa Aesar Supplier No. 041508.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 68478-92-2 | C8 H18 O Pt Si2 | 474.68 g/mol

Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst.Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, soln. in vinyl terminated polydimethylsiloxane is used as a catalyst in hydrosilylation reaction for the preparation of trans-beta-silanes and in reductive amination reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Catalyst for Si-H addition to olefins silicone vinyl addition room temperature cure catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68478-92-2
Molecular Formula C8 H18 O Pt Si2
Molecular Weight (g/mol) 474.68
MDL Number MFCD00151662
InChI Key RCNRJBWHLARWRP-UHFFFAOYSA-N
Synonym platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane, 1,1,3,3-tetramethyl-1,3-divinyldisiloxane, platinum salt, platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex solution, ksc628a5r, platinum 1,3-divinyl-1,1,3,3-tetramethyldisiloxane, dimethyl vinyl silyl oxy-dimethyl-vinyl-silane; platinum, ethenyl ethenyldimethylsilyl oxy dimethylsilane platinum, platinum 0-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, platinum,1,3-diethenyl-1,1,3,3-tetramethyldisiloxane complexes
PubChem CID 10959889
SMILES [Pt].C[Si](C)(O[Si](C)(C)C=C)C=C

Specifications

Color Orange
Density 0.984 g/mL
Flash Point >110°C (230°F)
Quantity 5 g
Sensitivity Moisture sensitive
Solubility Information Miscible with toluene,xylene,isopropyl alcohol,double-vinyl silicone oil and vinyl terminated poly(dimethylsiloxane). Immiscible with water.
IUPAC Name ethenyl[(ethenyldimethylsilyl)oxy]dimethylsilane platinum
Formula Weight 474.68
Physical Form Liquid
Chemical Name or Material Platinum (0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane complex, Soln. in vinyl terminated polydimethylsiloxane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.