accessibility menu, dialog, popup

UserName

CH3NOHNOH3CCH3

CAS RN 6851-81-6

IUPAC Name: 1,5,5-trimethylimidazolidine-2,4-dione
Synonyms: 1,5,5-trimethylimidazolidine-2,4-dione
Molecular Weight (g/mol): 142.16
Molecular Formula: C6H10N2O2
InChi Key: ZNYIPTYJBRGSSL-UHFFFAOYSA-N
SMILES: CN1C(=O)NC(=O)C1(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results