missing translation for 'onlineSavingsMsg'
Learn More

1,5,5-Trimethylhydantoin, 98%

Product Code. 11441227
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441227 100 g 100g
11421227 5 g 5g
11431227 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441227 Supplier Thermo Scientific Alfa Aesar Supplier No. B24188.22
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6851-81-6 | C6H10N2O2 | 142.158 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6851-81-6
Molecular Formula C6H10N2O2
Molecular Weight (g/mol) 142.158
MDL Number MFCD00040439
InChI Key ZNYIPTYJBRGSSL-UHFFFAOYSA-N
Synonym 1,5,5-trimethylhydantoin, 3,4,4-trimethyl-2,5-dioxoimidazolidine, 1,5,5-trimethyl-imidazolidine-2,4-dione, 1,5,5-trimethyl-1,3-diazolidine-2,4-dione, hydantoin, 1,5,5-trimethyl, 2,4-imidazolidinedione, 1,5,5-trimethyl, 1,5,5 trimethylhydantoin, acmc-209o3u, 1,5,5-trimethyl-hydantoin, 1,5,5-trimethyl hydantoin
PubChem CID 81295
IUPAC Name 1,5,5-trimethylimidazolidine-2,4-dione
SMILES CC1(C(=O)NC(=O)N1C)C

Specifications

Melting Point 161°C to 164°C
Quantity 100 g
Formula Weight 142.16
Percent Purity 98%
Chemical Name or Material 1,5,5-Trimethylhydantoin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.