accessibility menu, dialog, popup

UserName

CAS RN 68584-22-5

H3CCH3SOHOO

CAS RN 68584-22-5

IUPAC Name: 4-(dodecan-3-yl)benzene-1-sulfonic acid
Synonyms: 4-(dodecan-3-yl)benzenesulfonic acid
Molecular Weight (g/mol): 326.5
Molecular Formula: C18H30O3S
InChi Key: QJRVOJKLQNSNDB-UHFFFAOYNA-N
SMILES: CCCCCCCCCC(CC)C1=CC=C(C=C1)S(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results