accessibility menu, dialog, popup

UserName

CH3H2NNH2H3C

CAS RN 6864-37-5

IUPAC Name: 4-[(4-amino-3-methylcyclohexyl)methyl]-2-methylcyclohexan-1-amine
Synonyms: 4-[(4-amino-3-methylcyclohexyl)methyl]-2-methylcyclohexan-1-amine
Molecular Weight (g/mol): 238.42
Molecular Formula: C15H30N2
InChi Key: IGSBHTZEJMPDSZ-UHFFFAOYNA-N
SMILES: CC1CC(CC2CCC(N)C(C)C2)CCC1N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results