accessibility menu, dialog, popup

UserName

NH2H2NLiHC

CAS RN 6867-30-7

IUPAC Name: lithium(1+) acetylene ethane-1,2-diamine
Synonyms: lithium(1+) acetylene ethylenediamine
Molecular Weight (g/mol): 92.07
Molecular Formula: C4H9LiN2
InChi Key: WMWSRIHFAVOHSW-UHFFFAOYSA-N
SMILES: [Li+].C#[C-].NCCN
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (3)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)