missing translation for 'onlineSavingsMsg'
Learn More

Lithium acetylide ethylenediamine complex, tech. 85%, Thermo Scientific Chemicals

Product Code. 11466780
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466780 100 g 100g
11456780 25 g 25g
11368785 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11466780 Supplier Thermo Scientific Alfa Aesar Supplier No. L12401.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 6867-30-7 | Molecular Formula: C4H9LiN2 | Molecular Weight: 92.07 g/mol

Lithium acetylide ethylenediamine complex is used in the synthesis of a chiral cyclopropanol. It is also used as an alkynylating reagent for the enantioselective alkynylation of ketones catalyzed by lithium binaphtholate without using other metal sources.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium acetylide ethylenediamine complex is used in the synthesis of a chiral cyclopropanol. It is also used as an alkynylating reagent for the enantioselective alkynylation of ketones catalyzed by lithium binaphtholate without using other metal sources.

Solubility
Soluble in ethylene diamine, n-propyl amine, n-butyl amine, pyridine, dimethyl formamaide, diglyme, dioxan and dimethylsulfoxide.

Notes
Moisture and light sensitive. Store in cool place. Incompatible with alcohols, acids, oxidizing agents, water. It reacts with water to form lithium hydroxide and acetylene, which is a flammable gas. Over time, pressure may increase causing containers to burst. Handle and open container with care.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6867-30-7
Molecular Formula C4H9LiN2
Molecular Weight (g/mol) 92.07
MDL Number MFCD00013183
InChI Key QJQWXTYPTBEPGS-UHFFFAOYSA-N
Synonym lithium acetylide ethylenediamine complex, lithium acetylide, ethylenediamine complex, lithium; ethane-1,2-diamine; ethyne, 1,2-ethanediamine compd. with lithium acetylide li c2h, lithium, 1,2-ethanediamine-kn1,kn2 ethynyl, 1,2-ethanediamine, compd. with lithium acetylide li c2h 1:?, c2h8n2.c2hli, lithium acetylide, compd. with 1,2-ethanediamine, lithium acetylide, ethylene diamine complex 1:1, lithium acetylide, compound with ethane-1,2-diamine
PubChem CID 2724010
IUPAC Name lithium;ethane-1,2-diamine;ethyne
SMILES [Li]C#C.NCCN

Specifications

Melting Point ∽76°C (decomposition)
Density 1.04
Odor Amine-like
Linear Formula HC≡CLi·NH2CH2CH2NH2
Quantity 100 g
UN Number UN3131
Beilstein 3593824
Sensitivity Moisture sensitive
Solubility Information Soluble in ethylene diamine,n-propyl amine,n-butyl amine,pyridine,dimethyl formamaide,diglyme,dioxan and dimethylsulfoxide.
Formula Weight 92.07
Percent Purity 90%
Grade Technical
Chemical Name or Material Lithium acetylide ethylenediamine complex
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.