accessibility menu, dialog, popup

UserName

H3CNSOH

CAS RN 68867-18-5

IUPAC Name: 2-methyl-1,3-benzothiazol-6-ol
Synonyms: 2-methyl-1,3-benzothiazol-6-ol
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: ROFBPPIQUBJMRO-UHFFFAOYSA-N
SMILES: CC1=NC2=CC=C(O)C=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results