accessibility menu, dialog, popup

UserName

NaH3CS

CAS RN 6898-84-6

IUPAC Name: sodium propylsulfanide
Synonyms: sodium propylsulfanide
Molecular Weight (g/mol): 98.14
Molecular Formula: C3H7NaS
InChi Key: XVSFHIIADLZQJP-UHFFFAOYSA-M
SMILES: [Na+].CCC[S-]
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)