accessibility menu, dialog, popup

UserName

HONNHOHO

CAS RN 690631-98-2

IUPAC Name: 3-(3-hydroxyphenyl)-1H-pyrazole-5-carboxylic acid
Synonyms: 5-(3-hydroxyphenyl)-2H-pyrazole-3-carboxylic acid
Molecular Weight (g/mol): 204.19
Molecular Formula: C10H8N2O3
InChi Key: KOWRVFWZCLHEIU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=NN1)C1=CC(O)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results