accessibility menu, dialog, popup

UserName

H3CNOOH3C

CAS RN 6919-61-5

IUPAC Name: N-methoxy-N-methylbenzamide
Synonyms: N-methoxy-N-methylbenzamide
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: UKERDACREYXSIV-UHFFFAOYSA-N
SMILES: CON(C)C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results