accessibility menu, dialog, popup

UserName

OONN

CAS RN 6925-00-4

IUPAC Name: quinoxaline-6-carboxylate
Synonyms: quinoxaline-6-carboxylate
Molecular Weight (g/mol): 173.15
Molecular Formula: C9H5N2O2
InChi Key: JGQDBVXRYDEWGM-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C2N=CC=NC2=C1
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results