missing translation for 'onlineSavingsMsg'
Learn More

6-Quinoxalinecarboxylic acid, 95%, Thermo Scientific™

Product Code. 10744383
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10744383 10 g Amber Glass Bottle 10g
10130732 1 g Amber Glass Bottle 1g
10707095 50 g Amber Glass Bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10744383 Supplier Thermo Scientific Maybridge Supplier No. CC11701EA
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 6925-00-4
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
InChI Key JGQDBVXRYDEWGM-UHFFFAOYSA-N
Synonym 6-quinoxalinecarboxylic acid, chinoxalin-6-carbonsaeure, quinoxaline-6-carboxylicacid, pubchem15419, quinoxaline-6-carboxylic acid
PubChem CID 674813
IUPAC Name quinoxaline-6-carboxylic acid
SMILES C1=CC2=NC=CN=C2C=C1C(=O)O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 174.16
Percent Purity 95%
Chemical Name or Material 6-Quinoxalinecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.