accessibility menu, dialog, popup

UserName

CAS RN 69355-99-3

O(R)HNOOCH3CH3CH3(S)H3CHOO

CAS RN 69355-99-3

IUPAC Name: 3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}butanoic acid
Synonyms: 3-(benzyloxy)-2-[(tert-butoxycarbonyl)amino]butanoic acid
Molecular Weight (g/mol): 309.36
Molecular Formula: C16H23NO5
InChi Key: CTXPLTPDOISPTE-UHFFFAOYNA-N
SMILES: CC(OCC1=CC=CC=C1)C(NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)