accessibility menu, dialog, popup

UserName

OHN

CAS RN 69395-13-7

IUPAC Name: 4-(2-hydroxyethyl)benzonitrile
Synonyms: 4-(2-hydroxyethyl)benzonitrile
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: RBSJBNYPTGMZIH-UHFFFAOYSA-N
SMILES: OCCC1=CC=C(C=C1)C#N
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)