missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyethyl)benzonitrile, 98%

Product Code. 11329677
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11329677 5 g 5g
11319677 250 mg 250mg
11309677 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11329677 Supplier Thermo Scientific Alfa Aesar Supplier No. 041283.06
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Organic intermediate | CAS: 69395-13-7 | C9H9NO | 147.177 g/mol

4-(2-Hydroxyethyl)benzonitrile is used to produce 4-(2-bromo-ethyl)-benzonitrile. It is used as organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69395-13-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00052461
InChI Key RBSJBNYPTGMZIH-UHFFFAOYSA-N
Synonym 4-2-hydroxyethyl benzonitrile, benzonitrile, 4-2-hydroxyethyl, 2-4-cyanophenyl ethanol, 2-4-cyanophenyl ethan-1-one, 4-2-hydroxyethyl benzenecarbonitrile, 4-cyanobenzeneethanol, p-cyanophenethylalcohol, p-cyanophenethyl alcohol, 4-cyanophenethyl alcohol, acmc-20a4x6
PubChem CID 2800815
IUPAC Name 4-(2-hydroxyethyl)benzonitrile
SMILES C1=CC(=CC=C1CCO)C#N

Specifications

Boiling Point 116°C to 118°C (0.5 mmHg)
Quantity 5 g
UN Number UN3439
Formula Weight 147.18
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material 4-(2-Hydroxyethyl)benzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.