accessibility menu, dialog, popup

UserName

SiH2

CAS RN 694-53-1

IUPAC Name: phenylsilyl
Synonyms: phenylsilane
Molecular Weight (g/mol): 107.21
Molecular Formula: C6H7Si
InChi Key: JAZKZOWBOMBYIX-UHFFFAOYSA-N
SMILES: [SiH2]C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results