missing translation for 'onlineSavingsMsg'
Learn More

Phenylsilane, TRC

Product Code. 30409890
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409890 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30409890 Supplier Toronto Research Chemicals Supplier No. TRCP33677525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phenylsilane
CAS 694-53-1
Molecular Formula C6H8Si
Quantity 25 g
Synonym Silyl-benzene, Monophenylsilane, NSC 179699, SIP 6750.0, Silylbenzene, Z 6047
InChI Formula InChI=1S/C6H8Si/c7-6-4-2-1-3-5-6/h1-5H,7H3
SMILES [SiH3]c1ccccc1
IUPAC Name phenylsilane
Molecular Weight (g/mol) 108.21
Formula Weight 108.0395
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.