accessibility menu, dialog, popup

UserName

CAS RN 6950-82-9

IUPAC Name: 2-(7-hydroxy-2-oxo-2H-chromen-4-yl)acetic acid
Synonyms: (7-hydroxy-2-oxochromen-4-yl)acetic acid
Molecular Weight (g/mol): 220.18
Molecular Formula: C11H8O5
InChi Key: BNHPMQBVNXMPDU-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(=O)OC2=CC(O)=CC=C12
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results