accessibility menu, dialog, popup

UserName

BrOPO

CAS RN 69891-92-5

IUPAC Name: [2-(1,3-dioxan-2-yl)ethyl]triphenylphosphanium bromide
Synonyms: [2-(1,3-dioxan-2-yl)ethyl]triphenylphosphanium bromide
Molecular Weight (g/mol): 457.35
Molecular Formula: C24H26BrO2P
InChi Key: XETDBHNHTOJWPZ-UHFFFAOYSA-M
SMILES: [Br-].C(C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C1OCCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results