CAS RN 69912-75-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4'-O-Methylcatechin, TRC
CAS: 69912-75-0 Molecular Formula: C16H16O6 Molecular Weight (g/mol): 304.29 Synonym: (2R,3S)-3,4-Dihydro-2-(3-hydroxy-4-methoxyphenyl)-2H-1-benzopyran-3,5,7-triol,(2R-trans)-3,4-Dihydro-2-(3-hydroxy-4-methoxyphenyl)-2H-1-benzopyran-3,5,7-triol,4'-O-Methyl-(+)-catechin; IUPAC Name: (2R,3S)-2-(3-hydroxy-4-methoxyphenyl)-3,4-dihydro-2H-chromene-3,5,7-triol SMILES: COc1ccc(cc1O)[C@H]2Oc3cc(O)cc(O)c3C[C@@H]2O