accessibility menu, dialog, popup

UserName

ONOClF

CAS RN 700-37-8

IUPAC Name: 4-chloro-2-fluoro-1-nitrobenzene
Synonyms: 4-chloro-2-fluoro-1-nitrobenzene
Molecular Weight (g/mol): 175.54
Molecular Formula: C6H3ClFNO2
InChi Key: PTCPUGKKWNMITF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results