missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-2-fluoronitrobenzene, 98%, Thermo Scientific™

Product Code. 15505916 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15505916 25 g 25g
15595906 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15505916 Supplier Thermo Scientific Acros Supplier No. 455930250
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 700-37-8
Molecular Formula C6H3ClFNO2
Molecular Weight (g/mol) 175.54
MDL Number MFCD00042211
InChI Key PTCPUGKKWNMITF-UHFFFAOYSA-N
Synonym 4-chloro-2-fluoronitrobenzene, 2-fluoro-4-chloronitrobenzene, 1-chloro-3-fluoro-4-nitrobenzene, benzene, 4-chloro-2-fluoro-1-nitro, 4-chloro-2-fluoro-1-nitro-benzene, 4-chlor-2-fluor-1-nitrobenzol, pubchem1051, acmc-1bl8e, 4-chloro2-fluoronitrobenzene, ksc380m1b
PubChem CID 69691
IUPAC Name 4-chloro-2-fluoro-1-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(Cl)C=C1F

Specifications

Melting Point 48.0°C to 50.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Formula Weight 175.55
Percent Purity 98%
Chemical Name or Material 4-Chloro-2-fluoronitrobenzene
compliance-icons
Product Identifier
  • 4-Chloro-2-fluoronitrobenzene, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.