accessibility menu, dialog, popup

UserName

CHFFF

CAS RN 704-41-6

IUPAC Name: 1-ethynyl-2-(trifluoromethyl)benzene
Synonyms: 1-ethynyl-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 170.13
Molecular Formula: C9H5F3
InChi Key: OOZKONVIIMFOKW-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1C#C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results