accessibility menu, dialog, popup

UserName

CH3NOCH3NCH3SSNOCH3NCH3NON(Z)

CAS RN 7064-31-5

IUPAC Name: 5-{[(5Z)-3-(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene]methyl}-4-methyl-6-(3-methylpiperidin-1-yl)-2-oxo-1-propyl-1,2-dihydropyridine-3-carbonitrile
Synonyms: 5-{[(5Z)-3-(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene]methyl}-4-methyl-6-(3-methylpiperidin-1-yl)-2-oxo-1-propylpyridine-3-carbonitrile
Molecular Weight (g/mol): 602.77
Molecular Formula: C31H34N6O3S2
InChi Key: LQPROPLOIBHLEY-XYGWBWBKNA-N
SMILES: CCCN1C(=O)C(C#N)=C(C)C(\C=C2/SC(=S)N(C2=O)C2=C(C)N(C)N(C2=O)C2=CC=CC=C2)=C1N1CCCC(C)C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results