accessibility menu, dialog, popup

UserName

H3CNHHNCH3HClHCl

CAS RN 70708-33-7

IUPAC Name: N1,N2-dimethylcyclohexane-1,2-diamine dihydrochloride
Synonyms: N1,N2-dimethylcyclohexane-1,2-diamine dihydrochloride
Molecular Weight (g/mol): 215.16
Molecular Formula: C8H20Cl2N2
InChi Key: UDYXZFBJGBDHHP-UHFFFAOYNA-N
SMILES: Cl.Cl.CNC1CCCCC1NC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results