missing translation for 'onlineSavingsMsg'
Learn More

trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride, 98%, Thermo Scientific™

Product Code. 15249706 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15249706 25 g 25g
15229706 1 g 1g
15239706 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15249706 Supplier Thermo Scientific Acros Supplier No. 451020250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 70708-33-7
Molecular Formula C8H18N2·2ClH
Molecular Weight (g/mol) 215.17
InChI Key UDYXZFBJGBDHHP-RHJRFJOKSA-N
Synonym 1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride, trans-n,n'-dimethyl-1,2-diaminocyclohexane dihydrochloride, 1,2-cyclohexanediamine, n,n'-dimethyl-, dihydrochloride, 1r-trans, trans-1r,2r---n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride, trans-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride, 1r,2r-1-n,2-n-dimethylcyclohexane-1,2-diamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine 2hcl, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine dihydrochloride, 1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride
PubChem CID 44890801
IUPAC Name (1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine;dihydrochloride
SMILES CNC1CCCCC1NC.Cl.Cl

Specifications

Assay Percent Range 97.5 to max. 102.5% min. (Argentometry)
Quantity 25 g
Formula Weight 215.17
Percent Purity 98%
Chemical Name or Material trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride
compliance-icons
Product Identifier
  • trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.