CAS RN 70757-34-5
CAS RN 70757-34-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Keto Diclofenac Sodium Salt, TRC
CAS: 70757-34-5 Molecular Formula: C14 H8 Cl2 N O3 . Na Molecular Weight (g/mol): 332.11 Synonym: 2-[(2,6-Dichlorophenyl)amino]-α-oxo-benzeneacetic Acid Sodium Salt IUPAC Name: sodium;2-[2-(2,6-dichloroanilino)phenyl]-2-oxoacetate SMILES: [Na+].[O-]C(=O)C(=O)c1ccccc1Nc2c(Cl)cccc2Cl