missing translation for 'onlineSavingsMsg'
Learn More

Keto Diclofenac Sodium Salt, TRC

Product Code. 30399474
Change view
Click to view available options
Quantity:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399474 500 mg
30397167 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399474 Supplier Toronto Research Chemicals Supplier No. TRCK188000500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70757-34-5
Molecular Formula C14 H8 Cl2 N O3 . Na
Molecular Weight (g/mol) 332.11
Synonym 2-[(2,6-Dichlorophenyl)amino]-α-oxo-benzeneacetic Acid Sodium Salt
IUPAC Name sodium;2-[2-(2,6-dichloroanilino)phenyl]-2-oxoacetate
SMILES [Na+].[O-]C(=O)C(=O)c1ccccc1Nc2c(Cl)cccc2Cl

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C14H9Cl2NO3.Na/c15-9-5-3-6-10(16)12(9)17-11-7-2-1-4-8(11)13(18)14(19)20;/h1-7,17H,(H,19,20);/q;+1/p-1
Formula Weight 330.9779
Percent Purity >95%
Chemical Name or Material Keto Diclofenac Sodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.