accessibility menu, dialog, popup

UserName

H3COOFFF

CAS RN 710-18-9

IUPAC Name: 1-methoxy-4-(trifluoromethoxy)benzene
Synonyms: 1-methoxy-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 192.14
Molecular Formula: C8H7F3O2
InChi Key: NOAFZIOGGDPYKK-UHFFFAOYSA-N
SMILES: COC1=CC=C(OC(F)(F)F)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results