accessibility menu, dialog, popup

UserName

CAS RN 71203-35-5

H3CONNSOOH2N

CAS RN 71203-35-5

IUPAC Name: 4-[5-(4-methoxyphenyl)-3-phenyl-4,5-dihydro-1H-pyrazol-1-yl]benzene-1-sulfonamide
Synonyms: 4-[5-(4-methoxyphenyl)-3-phenyl-4,5-dihydropyrazol-1-yl]benzenesulfonamide
Molecular Weight (g/mol): 407.49
Molecular Formula: C22H21N3O3S
InChi Key: QBNZBMVRFYREHK-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C1CC(=NN1C1=CC=C(C=C1)S(N)(=O)=O)C1=CC=CC=C1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results