CAS RN 713-36-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Benzyl-2-methylbenzene, TRC
CAS: 713-36-0 Molecular Formula: C14H14 Molecular Weight (g/mol): 182.26 Synonym: 1-Methyl-2-(phenylmethyl)benzene;(2-Methylphenyl)phenylmethane;2-Benzyltoluene;2-Methyldiphenylmethane;NSC 75366;Phenyl(o-methylphenyl)methane;Phenyl-2-tolylmethane;Phenyl-o-tolylmethane;o-Benzyltoluene;o-Methyldiphenylmethane;o-Tolylphenylmethane;alpha-Phenyl-o-xylene; IUPAC Name: 1-benzyl-2-methylbenzene SMILES: Cc1ccccc1Cc2ccccc2