accessibility menu, dialog, popup

UserName

CAS RN 71434-68-9

IUPAC Name: (6-chlorohexyl)benzene
Synonyms: (6-chlorohexyl)benzene
Molecular Weight (g/mol): 196.72
Molecular Formula: C12H17Cl
InChi Key: RCVNHNOEUMFFDY-UHFFFAOYSA-N
SMILES: ClCCCCCCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results